Learn More
Medchemexpress LLC HY-16023B 5mg Medchemexpress, (Rac)-Acolbifene CAS:151533-34-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY16023B5MG
Encompass_Preferred
Medchemexpress, HY-16023B 5mg (Rac)-Acolbifene CAS:151533-34-5 (Rac)-Acolbifene (EM-343; (Rac)-EM-652) is the racemic form of EM652 (estrogen receptor antagonist), has anti-estrogenic and estrogenic activities. (Rac)-Acolbifene (EM-343; (Rac)-EM-652) contains a piperidine ring, shows good pharmacological profile,relative binding affinity (RBA)=380. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1864