Learn More
Medchemexpress LLC (Rac)-AMG8379 ((Rac)-AMG8380) | 1641574-26-6 | 99.25% | 543.93 | 10 MG
SDP
Catalog No. 5000347149
Encompass_Preferred
(Rac)-AMG8379, also known as (Rac)-AMG8380, is a racemate of AMG8379. It is described as a potent, orally active, and selective sulfonamide antagonist of NaV1.7. It exhibits IC50 values of 8.5 nM for hNaV1.7 and 18.6 nM for mNaV1.7.
  • Potent NaV1.7 antagonist.
  • Orally active compound.
  • Selective sulfonamide.
  • Available in various quantities for research needs.
Catalog No. 50-003-47149 Supplier Medchemexpress LLC Supplier No. HY108425B10MG
(Rac)-AMG8379, also known as (Rac)-AMG8380, is a racemate of AMG8379. It is described as a potent, orally active, and selective sulfonamide antagonist of NaV1.7. It exhibits IC50 values of 8.5 nM for hNaV1.7 and 18.6 nM for mNaV1.7.
  • Potent NaV1.7 antagonist.
  • Orally active compound.
  • Selective sulfonamide.
  • Available in various quantities for research needs.