Learn More
TARGETMOL CHEMICALS INC Rac-AMXT-1501 4HCl 1MG
SDP
Catalog No. 502880791
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes.(Rac)-AMXT-1501 4HCl is a polyamine transport inhibitor that inhibits polyamine transport and acts synergistically with cisplatin in HNSCC. (Rac)-AMXT-1501 4HCl has potential antimicrobial activity and inhibits neuroblastoma cell proliferation and pneumococcal pod biosynthesis by targeting ornithine decarboxylase and polyamine transport. Purity 98.31%
Catalog No. 50-288-0791 Supplier TARGETMOL CHEMICALS INC Supplier No. T103131MG
Also available in 2 mg, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes.(Rac)-AMXT-1501 4HCl is a polyamine transport inhibitor that inhibits polyamine transport and acts synergistically with cisplatin in HNSCC. (Rac)-AMXT-1501 4HCl has potential antimicrobial activity and inhibits neuroblastoma cell proliferation and pneumococcal pod biosynthesis by targeting ornithine decarboxylase and polyamine transport. Purity 98.31%