Learn More
Medchemexpress LLC (Rac)-Apremilast | 253168-86-4 | 99.5% | 460.50 | 50 MG
SDP
Catalog No. 5000429234
Encompass_Preferred
(Rac)-Apremilast ((Rac)-CC-10004) is the racemic mixture of Apremilast. It possesses anti-inflammatory properties and is utilized for research concerning inflammatory diseases, such as psoriasis.
  • Racemic mixture of Apremilast.
  • Exhibits anti-inflammatory properties.
  • Useful for research on inflammatory diseases like psoriasis.
  • Inhibits TNFalpha production in LPS-stimulated human PBMC with an IC50 of 0.19 μM.
  • IC50 for hPDE4D is 0.3 nM.
Catalog No. 50-004-29234 Supplier Medchemexpress LLC Supplier No. HYI063550MG
(Rac)-Apremilast ((Rac)-CC-10004) is the racemic mixture of Apremilast. It possesses anti-inflammatory properties and is utilized for research concerning inflammatory diseases, such as psoriasis.
  • Racemic mixture of Apremilast.
  • Exhibits anti-inflammatory properties.
  • Useful for research on inflammatory diseases like psoriasis.
  • Inhibits TNFalpha production in LPS-stimulated human PBMC with an IC50 of 0.19 μM.
  • IC50 for hPDE4D is 0.3 nM.