Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Rac)-apremilast | 253168-86-4 | 99.5% | 460.50 g·mol⁻¹ | C22H24N2O7S | 10 MG
SDP

Catalog No. 5000297602
Encompass_Preferred

Racemic apremilast is the racemate of apremilast, a small-molecule phosphodiesterase 4 (PDE4) inhibitor used for anti-inflammatory research. Supplied as a pre-weighed 10 mg solid, this research-grade reagent is suitable for biochemical assays, analytical reference standards, and method development.

  • Racemic mixture of apremilast for research applications.
  • PDE4 inhibitor used to study inflammatory signaling pathways.
  • High purity (reported ~99.5%).
  • Molecular weight: 460.50 g·mol⁻¹.
  • CAS number: 253168-86-4.
  • Supplied as a 10 mg pre-weighed sample for analytical use.

Catalog No. 50-002-97602 Supplier Medchemexpress LLC Supplier No. HYI063510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Racemic apremilast is the racemate of apremilast, a small-molecule phosphodiesterase 4 (PDE4) inhibitor used for anti-inflammatory research. Supplied as a pre-weighed 10 mg solid, this research-grade reagent is suitable for biochemical assays, analytical reference standards, and method development.

  • Racemic mixture of apremilast for research applications.
  • PDE4 inhibitor used to study inflammatory signaling pathways.
  • High purity (reported ~99.5%).
  • Molecular weight: 460.50 g·mol⁻¹.
  • CAS number: 253168-86-4.
  • Supplied as a 10 mg pre-weighed sample for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.