Learn More
Cayman Chemical rac-Aprepitant-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2748471-42-1, 1 mg
SDP
Catalog No. 502932477
Encompass_Preferred
An internal standard for the quantification of aprepitant by GC- or LC-MS
Catalog No. 50-293-2477 Supplier Cayman Chemical Supplier No. 252321 MG
An internal standard for the quantification of aprepitant by GC- or LC-MS