Learn More
Cayman Chemical rac-AprepItnt-d4 500ug
SDP
Catalog No. 502932516
Encompass_Preferred
An internal standard for the quantification of aprepitant by GC- or LC-MS
Catalog No. 50-293-2516 Supplier Cayman Chemical Supplier No. 25232500 UG
An internal standard for the quantification of aprepitant by GC- or LC-MS