Learn More
Medchemexpress LLC (rac)-AR-13503 ((rac)-AR-13324 M1 metabolite) | 1254032-16-0 | 99.1% | 321.37 g/mol | C19H19N3O2 | 5MG
SDP
Catalog No. 5000238126
Encompass_Preferred
(rac)-AR-13503 is a small-molecule ROCK/PKC inhibitor and an active metabolite of netarsudil. It inhibits angiogenesis, enhances retinal pigment epithelium (RPE) barrier function, and reduces aberrant neovascularization in preclinical models. The compound is provided as a high-purity research-grade solid for in vitro and in vivo studies.
  • Inhibits ROCK and PKC signaling.
  • Reduces angiogenesis and aberrant neovascularization in animal models.
  • Enhances retinal pigment epithelium barrier function in dose-dependent assays.
  • Active in vitro with HUVEC tube formation IC50 of approximately 21 nM.
  • Provided as a high-purity research compound suitable for biological assays.
  • Available in multiple small-mass package sizes for laboratory use.
Catalog No. 50-002-38126 Supplier Medchemexpress LLC Supplier No. HY1256395MG
(rac)-AR-13503 is a small-molecule ROCK/PKC inhibitor and an active metabolite of netarsudil. It inhibits angiogenesis, enhances retinal pigment epithelium (RPE) barrier function, and reduces aberrant neovascularization in preclinical models. The compound is provided as a high-purity research-grade solid for in vitro and in vivo studies.
  • Inhibits ROCK and PKC signaling.
  • Reduces angiogenesis and aberrant neovascularization in animal models.
  • Enhances retinal pigment epithelium barrier function in dose-dependent assays.
  • Active in vitro with HUVEC tube formation IC50 of approximately 21 nM.
  • Provided as a high-purity research compound suitable for biological assays.
  • Available in multiple small-mass package sizes for laboratory use.