Learn More
Medchemexpress LLC Rac-atomoxetine-d5 hydrochloride | 1398065-95-6 | MFCD04972577 | 99.0% | 296.85 | C17H17D5ClNO | 1 MG
SDP
Catalog No. 5000241474
Encompass_Preferred
(Rac)-Atomoxetine-d5 hydrochloride is the deuterium-labeled form of atomoxetine hydrochloride provided for research and analytical use. It serves as an isotopically labeled internal standard or tracer for quantitative assays, particularly mass spectrometry, and is supplied as the hydrochloride salt with molecular weight 296.85 and formula C17H17D5ClNO.
  • Deuterium-labeled internal standard for quantitative mass spectrometry.
  • Hydrochloride salt form suitable for analytical workflows.
  • High chemical purity (99.0%).
  • Supplied in small, accurately weighed quantities for trace analysis.
  • Suitable as tracer and internal standard for LC-MS/MS and GC-MS assays.
Catalog No. 50-002-41474 Supplier Medchemexpress LLC Supplier No. HY107370AS11MG
(Rac)-Atomoxetine-d5 hydrochloride is the deuterium-labeled form of atomoxetine hydrochloride provided for research and analytical use. It serves as an isotopically labeled internal standard or tracer for quantitative assays, particularly mass spectrometry, and is supplied as the hydrochloride salt with molecular weight 296.85 and formula C17H17D5ClNO.
  • Deuterium-labeled internal standard for quantitative mass spectrometry.
  • Hydrochloride salt form suitable for analytical workflows.
  • High chemical purity (99.0%).
  • Supplied in small, accurately weighed quantities for trace analysis.
  • Suitable as tracer and internal standard for LC-MS/MS and GC-MS assays.