Learn More
Medchemexpress LLC (Rac)-atropine-d3 | 1276197-36-4 | 99.1% | 292.39 g/mol | C17H20D3NO3 | 10 MG
SDP
Catalog No. 5000297748
Encompass_Preferred
(Rac)-Atropine-d3 is the deuterium-labeled form of racemic atropine intended for use as a tracer or internal standard in analytical methods such as NMR, GC-MS, and LC-MS. It is supplied as a white to off-white solid and is offered in milligram-scale research pack sizes.
  • Deuterium-labeled racemic atropine for analytical use.
  • Suitable as tracer or internal standard for NMR, GC-MS, and LC-MS.
  • High reported purity (≈99.1%).
  • White to off-white solid, convenient for handling and storage.
  • Recommended storage: powder at -20°C for long-term stability.
  • Available in milligram-scale quantities for method development and quantitation.
Catalog No. 50-002-97748 Supplier Medchemexpress LLC Supplier No. HYB1205S10MG
(Rac)-Atropine-d3 is the deuterium-labeled form of racemic atropine intended for use as a tracer or internal standard in analytical methods such as NMR, GC-MS, and LC-MS. It is supplied as a white to off-white solid and is offered in milligram-scale research pack sizes.
  • Deuterium-labeled racemic atropine for analytical use.
  • Suitable as tracer or internal standard for NMR, GC-MS, and LC-MS.
  • High reported purity (≈99.1%).
  • White to off-white solid, convenient for handling and storage.
  • Recommended storage: powder at -20°C for long-term stability.
  • Available in milligram-scale quantities for method development and quantitation.