Learn More
Medchemexpress LLC (Rac)-Atropine-d3 | 1276197-36-4 | 99.1% | 5 MG
SDP
Catalog No. 5000411294
Encompass_Preferred
(Rac)-Atropine-d3 is the deuterium labeled (Rac)-Atropine, designed for research use only. It serves as a tracer and internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. Deuteration, the substitution of deuterium for hydrogen, is recognized for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer in analytical studies
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, and LC-MS applications
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-11294 Supplier Medchemexpress LLC Supplier No. HYB1205S5MG
(Rac)-Atropine-d3 is the deuterium labeled (Rac)-Atropine, designed for research use only. It serves as a tracer and internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. Deuteration, the substitution of deuterium for hydrogen, is recognized for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer in analytical studies
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, and LC-MS applications
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs