Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Rac)-AZD 6482 | 663620-70-0 | 98.5% | 408.45 g·mol⁻¹ | C22H24N4O4 | 10 MG
SDP

Catalog No. 5000298187
Encompass_Preferred

(Rac)-AZD 6482 is the racemate of AZD 6482, a potent and selective p110β (PI3Kβ) inhibitor with an IC50 of 0.69 nM. The compound is supplied for research use and includes guidance for solvent preparation, storage, and in vivo formulation.

  • Racemic p110β inhibitor with sub-nanomolar potency (IC50 0.69 nM).
  • Molecular formula C22H24N4O4 and molecular weight 408.45 g·mol⁻¹.
  • Recommended solvent: DMSO; detailed stock preparation volumes provided.
  • Available pack sizes for research: 1 mg, 5 mg, 10 mg.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solution -80°C (6 months) or -20°C (1 month).
  • Reported HPLC purity approximately 98.5%.

Catalog No. 50-002-98187 Supplier Medchemexpress LLC Supplier No. HY7512410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rac)-AZD 6482 is the racemate of AZD 6482, a potent and selective p110β (PI3Kβ) inhibitor with an IC50 of 0.69 nM. The compound is supplied for research use and includes guidance for solvent preparation, storage, and in vivo formulation.

  • Racemic p110β inhibitor with sub-nanomolar potency (IC50 0.69 nM).
  • Molecular formula C22H24N4O4 and molecular weight 408.45 g·mol⁻¹.
  • Recommended solvent: DMSO; detailed stock preparation volumes provided.
  • Available pack sizes for research: 1 mg, 5 mg, 10 mg.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solution -80°C (6 months) or -20°C (1 month).
  • Reported HPLC purity approximately 98.5%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.