Learn More
Medchemexpress LLC (Rac)-AZD 6482 | 663620-70-0 | 98.5% | C22H24N4O4 | 5 MG
SDP
Catalog No. 5000433672
Encompass_Preferred
(Rac)-AZD 6482 is the racemate of AZD 6482. It is a potent and selective p110β inhibitor, with an IC50 of 0.69 nM. This compound is intended for research use only.
  • Potent and selective p110β inhibitor
  • IC50 of 0.69 nM
  • Racemate of AZD 6482
  • Solid appearance
  • White to light yellow color
  • For research use only
Catalog No. 50-004-33672 Supplier Medchemexpress LLC Supplier No. HY751245MG
(Rac)-AZD 6482 is the racemate of AZD 6482. It is a potent and selective p110β inhibitor, with an IC50 of 0.69 nM. This compound is intended for research use only.
  • Potent and selective p110β inhibitor
  • IC50 of 0.69 nM
  • Racemate of AZD 6482
  • Solid appearance
  • White to light yellow color
  • For research use only