Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Rac)-Baxdrostat | 1428652-15-6 | 99.3% | C22H25N3O2 | 10MG
SDP

Catalog No. 5000258902
Encompass_Preferred

(Rac)-Baxdrostat is the racemic isomer of Baxdrostat, a selective aldosterone synthase (CYP11B2) inhibitor. Supplied as a research reagent, the racemate is intended for use as an experimental control in studies of aldosterone biosynthesis, mineralocorticoid pathway modulation, and assay validation.

  • Racemic isomer suitable as an experimental control.
  • Useful for studies of aldosterone synthase activity and mineralocorticoid signaling.
  • Molecular formula C22H25N3O2; molecular weight 363.45 g·mol⁻¹.
  • Provided as a solid suitable for formulation and analytical preparation.

Catalog No. 50-002-58902 Supplier Medchemexpress LLC Supplier No. HY132809B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rac)-Baxdrostat is the racemic isomer of Baxdrostat, a selective aldosterone synthase (CYP11B2) inhibitor. Supplied as a research reagent, the racemate is intended for use as an experimental control in studies of aldosterone biosynthesis, mineralocorticoid pathway modulation, and assay validation.

  • Racemic isomer suitable as an experimental control.
  • Useful for studies of aldosterone synthase activity and mineralocorticoid signaling.
  • Molecular formula C22H25N3O2; molecular weight 363.45 g·mol⁻¹.
  • Provided as a solid suitable for formulation and analytical preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.