Learn More
Medchemexpress LLC HY-112316B 10mg Medchemexpress, (Rac)-BAY1238097 CAS:1564268-19-4 Purity:>98%
SDP
Catalog No. 501969082
Encompass_Preferred
Medchemexpress, HY-112316B 10mg (Rac)-BAY1238097 CAS:1564268-19-4 (Rac)-BAY1238097 is a BET inhibitor, with an IC50 of 1.02 μM for BRD4. Used in cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9082 Supplier Medchemexpress LLC Supplier No. HY112316B10MG
Medchemexpress, HY-112316B 10mg (Rac)-BAY1238097 CAS:1564268-19-4 (Rac)-BAY1238097 is a BET inhibitor, with an IC50 of 1.02 μM for BRD4. Used in cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.