Learn More
Medchemexpress LLC (Rac)-BAY1238097 | 1564268-19-4 | 99.7% | 451.56 | 10 MM 1 ML
SDP
Catalog No. 5000438136
Encompass_Preferred
(Rac)-BAY1238097 is a BET inhibitor, with an IC50 of 1.02 μM for BRD4. It is used in cancer research.
  • BET inhibitor
  • IC50 of 1.02 μM for BRD4
  • Used in cancer research
Catalog No. 50-004-38136 Supplier Medchemexpress LLC Supplier No. HY112316B10MM/1ML
(Rac)-BAY1238097 is a BET inhibitor, with an IC50 of 1.02 μM for BRD4. It is used in cancer research.
  • BET inhibitor
  • IC50 of 1.02 μM for BRD4
  • Used in cancer research