Learn More
Medchemexpress LLC (Rac)-BAY1238097 | 1564268-19-4 | 99.7% | 451.56 | 100 MG
SDP
Catalog No. 5000438264
Encompass_Preferred
(Rac)-BAY1238097 is a BET inhibitor that has an IC50 of 1.02 μM for BRD4 and is used in cancer research.
  • Functions as a BET inhibitor
  • Exhibits an IC50 of 1.02 μM for BRD4
  • Utilized in cancer research
  • Active in epigenetic reader domain
  • Relevant for cancer and blood-related diseases
Catalog No. 50-004-38264 Supplier Medchemexpress LLC Supplier No. HY112316B100MG
(Rac)-BAY1238097 is a BET inhibitor that has an IC50 of 1.02 μM for BRD4 and is used in cancer research.
  • Functions as a BET inhibitor
  • Exhibits an IC50 of 1.02 μM for BRD4
  • Utilized in cancer research
  • Active in epigenetic reader domain
  • Relevant for cancer and blood-related diseases