Learn More
Medchemexpress LLC (Rac)-BAY1238097 | 1564268-19-4 | 99.7% | 451.56 | 50 MG
SDP
Catalog No. 5000438644
Encompass_Preferred
(Rac)-BAY1238097 is a BET inhibitor with an IC50 of 1.02 μM for BRD4. It is used in cancer research.
  • Used in cancer research.
  • Solid form, light yellow to yellow color.
Catalog No. 50-004-38644 Supplier Medchemexpress LLC Supplier No. HY112316B50MG
(Rac)-BAY1238097 is a BET inhibitor with an IC50 of 1.02 μM for BRD4. It is used in cancer research.
  • Used in cancer research.
  • Solid form, light yellow to yellow color.