Learn More
Medchemexpress LLC (Rac)-Bedaquiline-d6 | 2517573-53-2 | 99.0% | 561.54 | 1 MG
SDP
Catalog No. 5000361484
Encompass_Preferred
(Rac)-Bedaquiline-d6 is a deuterium-labeled version of Bedaquiline that inhibits *Mycobacterium tuberculosis* F1FO-ATP synthase. It exhibits uncoupler activity and is used for multi-agent resistant tuberculosis. Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-61484 Supplier Medchemexpress LLC Supplier No. HY14881S11MG
(Rac)-Bedaquiline-d6 is a deuterium-labeled version of Bedaquiline that inhibits *Mycobacterium tuberculosis* F1FO-ATP synthase. It exhibits uncoupler activity and is used for multi-agent resistant tuberculosis. Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS