Learn More
Medchemexpress LLC (Rac)-Bedaquiline-d6 | 2517573-53-2 | 99.0% | 561.54 | 5 MG
SDP
Catalog No. 5000362363
Encompass_Preferred
(Rac)-Bedaquiline-d6 is the deuterium labeled Bedaquiline. It is a diarylquinoline agent that inhibits *Mycobacterium tuberculosis* (Mtb) F1FO-ATP synthase by targeting both the c- and the ε-subunit. This compound exhibits uncoupler activity and is used for multi-agent resistant tuberculosis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-62363 Supplier Medchemexpress LLC Supplier No. HY14881S15MG
(Rac)-Bedaquiline-d6 is the deuterium labeled Bedaquiline. It is a diarylquinoline agent that inhibits *Mycobacterium tuberculosis* (Mtb) F1FO-ATP synthase by targeting both the c- and the ε-subunit. This compound exhibits uncoupler activity and is used for multi-agent resistant tuberculosis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS