Learn More
TARGETMOL CHEMICALS INC RAC-BELINOSTAT 50MG
SDP
Catalog No. 502581076
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rac-Belinostat (PX-105684) is a novel hydroxamic acid-type histone deacetylase (HDAC) inhibitor with antineoplastic activity. Belinostat targets HDAC enzymes thereby inhibiting tumor cell proliferation inducing apoptosis promoting cellular differentiation and inhibiting angiogenesis. This agent may sensitize drug-resistant tumor cells to other antineoplastic agents possibly through a mechanism involving the down-regulation of thymidylate synthase. purity: 99%
Catalog No. 50-258-1076 Supplier TARGETMOL CHEMICALS INC Supplier No. T185250MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rac-Belinostat (PX-105684) is a novel hydroxamic acid-type histone deacetylase (HDAC) inhibitor with antineoplastic activity. Belinostat targets HDAC enzymes thereby inhibiting tumor cell proliferation inducing apoptosis promoting cellular differentiation and inhibiting angiogenesis. This agent may sensitize drug-resistant tumor cells to other antineoplastic agents possibly through a mechanism involving the down-regulation of thymidylate synthase. purity: 99%