Learn More
Cayman Chemical rac-Benpyrine, Purity: ≥98%, CAS: 1333714-43-4, 5 mg
SDP
Catalog No. 502913535
Encompass_Preferred
An inhibitor of the TNF-α-TNFR1 protein-protein interaction (IC50 = 0.109 µM); reduces TNF-α induced cytotoxicity in L-929 fibroblasts at 10 µM; prevents nuclear translocation of NF-κB in RAW 264.7 macrophages; improves survival in a mouse model of LPS-induced endotoxemia and decreases disease severity in mouse models of collagen-induced arthritis and imiquimod-induced psoriasis at 25 mg/kg
Catalog No. 50-291-3535 Supplier Cayman Chemical Supplier No. 366805 MG
An inhibitor of the TNF-α-TNFR1 protein-protein interaction (IC50 = 0.109 µM); reduces TNF-α induced cytotoxicity in L-929 fibroblasts at 10 µM; prevents nuclear translocation of NF-κB in RAW 264.7 macrophages; improves survival in a mouse model of LPS-induced endotoxemia and decreases disease severity in mouse models of collagen-induced arthritis and imiquimod-induced psoriasis at 25 mg/kg