Learn More
TARGETMOL CHEMICALS INC Rac-Benpyrine 5MG
SDP
Catalog No. 502879300
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. (Rac)-Benpyrine is a potent and orally active inhibitor of TNF-alpha. Purity 98.55%
Catalog No. 50-287-9300 Supplier TARGETMOL CHEMICALS INC Supplier No. T86925MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. (Rac)-Benpyrine is a potent and orally active inhibitor of TNF-alpha. Purity 98.55%