Learn More
Medchemexpress LLC (Rac)-Bepotastine besilate | 1415692-17-9 | 99.5% | C27H31ClN2O6S | 50 MG
SDP
Catalog No. 5000433306
Encompass_Preferred
(Rac)-Bepotastine besilate is an isomer of Bepotastine besilate (HY-A0015) and serves as an experimental control. Bepotastine besilate is a selective, orally active second-generation histamine H1 receptor antagonist that suppresses the expression of nerve growth factor (NGF). It has potential for research related to allergic rhinitis, allergic conjunctivitis, and urticaria/pruritus.
  • Isomer of Bepotastine besilate (HY-A0015)
  • Used as an experimental control
  • Selective and orally active second-generation histamine H1 receptor antagonist
  • Suppresses nerve growth factor (NGF) expression
  • Potential for research in allergic rhinitis, allergic conjunctivitis, and urticaria/pruritus
Catalog No. 50-004-33306 Supplier Medchemexpress LLC Supplier No. HYA0015A50MG
(Rac)-Bepotastine besilate is an isomer of Bepotastine besilate (HY-A0015) and serves as an experimental control. Bepotastine besilate is a selective, orally active second-generation histamine H1 receptor antagonist that suppresses the expression of nerve growth factor (NGF). It has potential for research related to allergic rhinitis, allergic conjunctivitis, and urticaria/pruritus.
  • Isomer of Bepotastine besilate (HY-A0015)
  • Used as an experimental control
  • Selective and orally active second-generation histamine H1 receptor antagonist
  • Suppresses nerve growth factor (NGF) expression
  • Potential for research in allergic rhinitis, allergic conjunctivitis, and urticaria/pruritus