Learn More
TARGETMOL CHEMICALS INC RAC-BHFF 25MG
SDP
Catalog No. 502579933
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. rac-BHFF ((RS)-BHFF) is potent and selective GABA B receptor positive allosteric modulator that increases the potency and efficacy of GABA ( > 15-fold and > 149% respectively). It exhibits anxiolytic activity in vivo and is orally active. purity: 98%
Catalog No. 50-257-9933 Supplier TARGETMOL CHEMICALS INC Supplier No. T417625MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. rac-BHFF ((RS)-BHFF) is potent and selective GABA B receptor positive allosteric modulator that increases the potency and efficacy of GABA ( > 15-fold and > 149% respectively). It exhibits anxiolytic activity in vivo and is orally active. purity: 98%