Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Rac-Binap

Catalog No. 481084500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-108-4500G 500 g
48-108-4100G 100 g
48-108-425G 25 g
48-108-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 48-108-4500G Supplier Sigma Aldrich Supplier No. 481084500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Rac-Binap

Specifications

CAS 98327-87-8
Melting Point 283°C to 286°C
Molecular Formula C44H32P2
Linear Formula [(C6H5)2PC10H6-]2
MDL Number MFCD00010805
Quantity 500 g
Synonym (+/-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene; 2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene; (+/-)-BINAP; [1,1'-Binaphthalene]-2,2'-diylbis[diphenylphosphine]
Molecular Weight (g/mol) 622.67 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.