Learn More
Medchemexpress LLC (Rac)-bio8898 | 402564-79-8 | 98.0% | 10 MG
SDP
Catalog No. 5000230689
Encompass_Preferred
(Rac)-BIO8898 is a synthetic small-molecule inhibitor of the CD40-CD154 (CD40L) protein-protein interaction, reported to inhibit soluble CD40L binding to CD40-Ig with an IC50 of approximately 25 μM. Supplied as a high-purity solid for research use.
  • Inhibits CD40-CD154 interaction; IC50 ≈ 25 μM.
  • Chemical formula C53H64N8O6; molecular weight 909.13 g·mol⁻1.
  • High purity (98.03%) suitable for biochemical and cell-based assays.
  • Available in small research sizes; soluble in DMSO.
  • For research use only; not for human or diagnostic use.
Catalog No. 50-002-30689 Supplier Medchemexpress LLC Supplier No. HY12266310MG
(Rac)-BIO8898 is a synthetic small-molecule inhibitor of the CD40-CD154 (CD40L) protein-protein interaction, reported to inhibit soluble CD40L binding to CD40-Ig with an IC50 of approximately 25 μM. Supplied as a high-purity solid for research use.
  • Inhibits CD40-CD154 interaction; IC50 ≈ 25 μM.
  • Chemical formula C53H64N8O6; molecular weight 909.13 g·mol⁻1.
  • High purity (98.03%) suitable for biochemical and cell-based assays.
  • Available in small research sizes; soluble in DMSO.
  • For research use only; not for human or diagnostic use.