Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Rac)-BMS-816336 | 1009365-98-3 | 99.9% | 341.4 g/mol | C21H27NO3 | 50 MG
SDP

Catalog No. 5000245910
Encompass_Preferred

(Rac)-BMS-816336 is the racemic form of a small-molecule 11β-hydroxysteroid dehydrogenase type 1 (11β-HSD1) inhibitor used in biochemical and preclinical research. It is supplied as a high-purity research reagent suitable for in vitro and in vivo assays, metabolic stability studies, and assay development.

  • Racemic small-molecule 11β-HSD1 inhibitor.
  • Molecular formula C21H27NO3; molecular weight 341.4 g/mol.
  • CAS 1009365-98-3 for substance identification.
  • High purity (99.9%) for research applications.
  • Powder form, soluble in DMSO for assay preparation.
  • Recommended storage: powder at -20°C or 4°C; in solvent at -80°C.
  • Intended for biochemical assays and preclinical studies (research use only).

Catalog No. 50-002-45910 Supplier Medchemexpress LLC Supplier No. HY101930A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rac)-BMS-816336 is the racemic form of a small-molecule 11β-hydroxysteroid dehydrogenase type 1 (11β-HSD1) inhibitor used in biochemical and preclinical research. It is supplied as a high-purity research reagent suitable for in vitro and in vivo assays, metabolic stability studies, and assay development.

  • Racemic small-molecule 11β-HSD1 inhibitor.
  • Molecular formula C21H27NO3; molecular weight 341.4 g/mol.
  • CAS 1009365-98-3 for substance identification.
  • High purity (99.9%) for research applications.
  • Powder form, soluble in DMSO for assay preparation.
  • Recommended storage: powder at -20°C or 4°C; in solvent at -80°C.
  • Intended for biochemical assays and preclinical studies (research use only).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.