Learn More
Medchemexpress LLC (Rac)-BMS-816336 | 1009365-98-3 | 99.9% | 341.44 g/mol | C21H27NO3 | 10 MG
SDP
Catalog No. 5000245627
Encompass_Preferred
(Rac)-BMS-816336 is the racemic form of a potent 11β-hydroxysteroid dehydrogenase type 1 (11β-HSD1) inhibitor supplied as a solid research chemical. It is characterized by the formula C21H27NO3 and a molecular weight of 341.44 g/mol, and is intended for laboratory and preclinical research use only.
Catalog No. 50-002-45627 Supplier Medchemexpress LLC Supplier No. HY101930A10MG
(Rac)-BMS-816336 is the racemic form of a potent 11β-hydroxysteroid dehydrogenase type 1 (11β-HSD1) inhibitor supplied as a solid research chemical. It is characterized by the formula C21H27NO3 and a molecular weight of 341.44 g/mol, and is intended for laboratory and preclinical research use only.