Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rac-brassinazole | 224047-41-0 | 5 MG
SDP

Catalog No. 5000232151
Encompass_Preferred

Rac-brassinazole is a triazole-type inhibitor of brassinosteroid biosynthesis that targets CYP90B and is used in research to modulate BR signaling pathways.

  • Triazole-type brassinosteroid biosynthesis inhibitor.
  • CAS 224047-41-0.
  • Molecular formula C18H18ClN3O.
  • Molecular weight 327.81 g/mol.
  • Appearance: solid.
  • Solubility: DMSO 50 mg/mL; in vivo formulations include DMSO/PEG300/Tween-80/saline and DMSO/corn oil.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-32151 Supplier Medchemexpress LLC Supplier No. HY1211615MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rac-brassinazole is a triazole-type inhibitor of brassinosteroid biosynthesis that targets CYP90B and is used in research to modulate BR signaling pathways.

  • Triazole-type brassinosteroid biosynthesis inhibitor.
  • CAS 224047-41-0.
  • Molecular formula C18H18ClN3O.
  • Molecular weight 327.81 g/mol.
  • Appearance: solid.
  • Solubility: DMSO 50 mg/mL; in vivo formulations include DMSO/PEG300/Tween-80/saline and DMSO/corn oil.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.