Learn More
Medchemexpress LLC (Rac)-Cemsidomide | 2504233-68-3 | 98.5% | C28H27N3O4 | 1 ML
SDP
Catalog No. 5000355890
Encompass_Preferred
(Rac)-Cemsidomide ((Rac)-CFT7455) is the racemate of Cemsidomide (HY-144841) with antitumor activity. Cemsidomide is a ubiquitin ligase pathway Ikaros/Aiolos degrader with molecular glue activity. It has an IC50 of 0.05 nM for NCIH929.1 cells.
  • Antitumor activity
  • Ubiquitin ligase pathway Ikaros/Aiolos degrader
  • Molecular glue activity
  • Targets Ikaros and Aiolos
  • For research use only
Catalog No. 50-003-55890 Supplier Medchemexpress LLC Supplier No. HY144841A10MM/1ML
(Rac)-Cemsidomide ((Rac)-CFT7455) is the racemate of Cemsidomide (HY-144841) with antitumor activity. Cemsidomide is a ubiquitin ligase pathway Ikaros/Aiolos degrader with molecular glue activity. It has an IC50 of 0.05 nM for NCIH929.1 cells.
  • Antitumor activity
  • Ubiquitin ligase pathway Ikaros/Aiolos degrader
  • Molecular glue activity
  • Targets Ikaros and Aiolos
  • For research use only