Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rac-clopidogrel-d3 sulfate | 2468372-74-7 | 422.92 g·mol⁻¹ | C16H15D3ClNO6S2 | 5 MG
SDP

Catalog No. 5000244509
Encompass_Preferred

(Rac)-Clopidogrel-d3 sulfate is a deuterium-labeled clopidogrel hydrogen sulfate intended for analytical and research use. It is designed to act as a stable isotope tracer and internal standard to support quantitative pharmacokinetic, metabolic, and bioanalytical studies.

  • Deuterium-labeled clopidogrel hydrogen sulfate for tracer and internal standard use.
  • Suitable for quantitative NMR, GC-MS, and LC-MS analyses.
  • Molecular weight approximately 422.92 g·mol⁻¹ and formula C16H15D3ClNO6S2.
  • Available in small quantities for analytical work (e.g., 5 MG).
  • Certificate of analysis and purity documentation available from the manufacturer.
  • Store and handle according to the certificate of analysis and datasheet recommendations.

Catalog No. 50-002-44509 Supplier Medchemexpress LLC Supplier No. HY107867S35MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rac)-Clopidogrel-d3 sulfate is a deuterium-labeled clopidogrel hydrogen sulfate intended for analytical and research use. It is designed to act as a stable isotope tracer and internal standard to support quantitative pharmacokinetic, metabolic, and bioanalytical studies.

  • Deuterium-labeled clopidogrel hydrogen sulfate for tracer and internal standard use.
  • Suitable for quantitative NMR, GC-MS, and LC-MS analyses.
  • Molecular weight approximately 422.92 g·mol⁻¹ and formula C16H15D3ClNO6S2.
  • Available in small quantities for analytical work (e.g., 5 MG).
  • Certificate of analysis and purity documentation available from the manufacturer.
  • Store and handle according to the certificate of analysis and datasheet recommendations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.