Learn More
Medchemexpress LLC (Rac)-Cotinine-d3 | 110952-70-0 | 99.0% | 1 MG
SDP
Catalog No. 5000413816
Encompass_Preferred
(Rac)-Cotinine-d3 is a deuterium-labeled form of (Rac)-Cotinine. This compound is used for research purposes, including as a tracer and an internal standard for quantitative analysis. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules as tracers during drug development, which can impact their pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs through deuteration
Catalog No. 50-004-13816 Supplier Medchemexpress LLC Supplier No. HYB1178S1MG
(Rac)-Cotinine-d3 is a deuterium-labeled form of (Rac)-Cotinine. This compound is used for research purposes, including as a tracer and an internal standard for quantitative analysis. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules as tracers during drug development, which can impact their pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules for quantitation
  • Potential to affect pharmacokinetic and metabolic profiles of drugs through deuteration