Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Rac)-Cotinine-d3 | 110952-70-0 | 99.9% | 179.23 | C10H9D3N2O | 10 MG
SDP

Catalog No. 5000296714
Encompass_Preferred

(Rac)-Cotinine-d3 is the deuterium-labeled racemic form of cotinine used as a stable isotope internal standard and tracer in analytical chemistry and bioanalysis. It is intended for use in quantitative mass spectrometry assays and metabolic studies where isotopically labeled standards improve accuracy and precision.

  • Deuterium-labeled internal standard for LC-MS/MS quantitation.
  • Racemic form compatible with assays targeting cotinine and metabolites.
  • High purity (99.9%) to minimize background interference.
  • Available in small milligram quantities for analytical workflows.
  • Molecular weight 179.23 and molecular formula C10H9D3N2O.

Catalog No. 50-002-96714 Supplier Medchemexpress LLC Supplier No. HYB1178S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rac)-Cotinine-d3 is the deuterium-labeled racemic form of cotinine used as a stable isotope internal standard and tracer in analytical chemistry and bioanalysis. It is intended for use in quantitative mass spectrometry assays and metabolic studies where isotopically labeled standards improve accuracy and precision.

  • Deuterium-labeled internal standard for LC-MS/MS quantitation.
  • Racemic form compatible with assays targeting cotinine and metabolites.
  • High purity (99.9%) to minimize background interference.
  • Available in small milligram quantities for analytical workflows.
  • Molecular weight 179.23 and molecular formula C10H9D3N2O.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.