Learn More
Medchemexpress LLC (Rac)-Cotinine-d3 | 110952-70-0 | 99.9% | 5 MG
SDP
Catalog No. 5000414058
Encompass_Preferred
(Rac)-Cotinine-d3 is the deuterium-labeled version of (Rac)-Cotinine. It is intended for research use only. The incorporation of stable heavy isotopes such as deuterium into drug molecules can serve as tracers for quantitative analysis during drug development, and has also been noted for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-14058 Supplier Medchemexpress LLC Supplier No. HYB1178S5MG
Add to Cart
Add to Cart
(Rac)-Cotinine-d3 is the deuterium-labeled version of (Rac)-Cotinine. It is intended for research use only. The incorporation of stable heavy isotopes such as deuterium into drug molecules can serve as tracers for quantitative analysis during drug development, and has also been noted for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.