Learn More
Medchemexpress LLC (Rac)-Cotinine-d3 | 110952-70-0 | 99.9% | 50 MG
SDP
Catalog No. 5000414057
Encompass_Preferred
(Rac)-Cotinine-d3 is the deuterium labeled (Rac)-Cotinine. Deuteration has gained attention due to its potential to impact the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-14057 Supplier Medchemexpress LLC Supplier No. HYB1178S50MG
(Rac)-Cotinine-d3 is the deuterium labeled (Rac)-Cotinine. Deuteration has gained attention due to its potential to impact the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.