Learn More
Medchemexpress LLC (Rac)-Cotinine-d4 | 350818-68-7 | 99.6% | 5 MG
SDP
Catalog No. 5000412876
Encompass_Preferred
(Rac)-Cotinine-d4 is the deuterium labeled (Rac)-Cotinine. It is for research use only and not sold to patients. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-12876 Supplier Medchemexpress LLC Supplier No. HYB1178S15MG
(Rac)-Cotinine-d4 is the deuterium labeled (Rac)-Cotinine. It is for research use only and not sold to patients. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.