Learn More
Medchemexpress LLC (Rac)-CP-601927 hydrochloride | 230615-01-7 | 99.7% | 263.69 | 25 MG
SDP
Catalog No. 5000352471
Encompass_Preferred
(Rac)-CP-601927 hydrochloride is the racemate of CP-601927, acting as a selective nAChR agonist. It demonstrates significant antidepressant-like effects in mice by targeting α4β2 and α3β4 nAChRs.
  • Selective nAChR agonist
  • Induces antidepressant-like effects in mice
  • Decreases cholinergic signaling
  • Targets α4β2 and α3β4 nAChRs
  • For research use only
Catalog No. 50-003-52471 Supplier Medchemexpress LLC Supplier No. 25MGHY138879A
(Rac)-CP-601927 hydrochloride is the racemate of CP-601927, acting as a selective nAChR agonist. It demonstrates significant antidepressant-like effects in mice by targeting α4β2 and α3β4 nAChRs.
  • Selective nAChR agonist
  • Induces antidepressant-like effects in mice
  • Decreases cholinergic signaling
  • Targets α4β2 and α3β4 nAChRs
  • For research use only