Learn More
Medchemexpress LLC (Rac)-dehydrocostus lactone | 74299-48-2 | 99.9% | 230.30 g/mol | C15H18O2 | 50 MG
SDP
Catalog No. 5000285070
Encompass_Preferred
(Rac)-Dehydrocostus lactone is a naturally occurring sesquiterpene lactone used in biochemical and pharmacological research. It has reported antiparasitic, anti-inflammatory, anticancer, antiviral, and antimicrobial activities and is used to study cytotoxicity and mechanism-of-action in vitro. The compound is supplied as a white to off-white solid with high reported purity and is soluble in DMSO for experimental use.
  • Naturally occurring sesquiterpene lactone.
  • Reported antiparasitic, anti-inflammatory, and anticancer activities.
  • High reported purity for experimental consistency.
  • Supplied as a white to off-white solid for ease of handling.
  • Soluble in DMSO at high concentrations for in vitro assays.
  • Suitable for biochemical and pharmacological research applications.
Catalog No. 50-002-85070 Supplier Medchemexpress LLC Supplier No. HYW39162550MG
(Rac)-Dehydrocostus lactone is a naturally occurring sesquiterpene lactone used in biochemical and pharmacological research. It has reported antiparasitic, anti-inflammatory, anticancer, antiviral, and antimicrobial activities and is used to study cytotoxicity and mechanism-of-action in vitro. The compound is supplied as a white to off-white solid with high reported purity and is soluble in DMSO for experimental use.
  • Naturally occurring sesquiterpene lactone.
  • Reported antiparasitic, anti-inflammatory, and anticancer activities.
  • High reported purity for experimental consistency.
  • Supplied as a white to off-white solid for ease of handling.
  • Soluble in DMSO at high concentrations for in vitro assays.
  • Suitable for biochemical and pharmacological research applications.