Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Rac)-dehydrocostus lactone | 74299-48-2 | MFCD00210277 | 99.9% | 230.30 g·mol⁻1 | C15H18O2 | 100 MG
SDP

Catalog No. 5000285155
Encompass_Preferred

(Rac)-Dehydrocostus lactone is a naturally occurring sesquiterpene lactone offered for research use. It exhibits reported anti-inflammatory, anticancer, antiviral, and antimicrobial activities and is intended as a high-purity analytical standard for in vitro and biochemical studies.

  • High purity: 99.9%.
  • Molecular formula: C15H18O2; molecular weight: 230.30 g·mol⁻1.
  • CAS number: 74299-48-2.
  • Supplied quantity: 100 MG.
  • Suitable as a reference standard for biological assays and analytical method development.
  • Includes datasheet with spectral and purity data.

Catalog No. 50-002-85155 Supplier Medchemexpress LLC Supplier No. HYW391625100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rac)-Dehydrocostus lactone is a naturally occurring sesquiterpene lactone offered for research use. It exhibits reported anti-inflammatory, anticancer, antiviral, and antimicrobial activities and is intended as a high-purity analytical standard for in vitro and biochemical studies.

  • High purity: 99.9%.
  • Molecular formula: C15H18O2; molecular weight: 230.30 g·mol⁻1.
  • CAS number: 74299-48-2.
  • Supplied quantity: 100 MG.
  • Suitable as a reference standard for biological assays and analytical method development.
  • Includes datasheet with spectral and purity data.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.