Learn More
Medchemexpress LLC (Rac)-EBET-1055 | 3031540-65-2 | 97.17% | 923.02 | 5 MG
SDP
Catalog No. 5000393175
Encompass_Preferred
(Rac)-EBET-1055 is the racemate of EBET-1055, a bromodomain and extra-terminal (BET) PROTAC degrader.
  • Effectively inhibits the growth of pancreatic ductal adenocarcinoma (PDAC).
  • Simultaneously modulates cancer-associated fibroblast (CAF) activity, upregulating all reporter gene activities in organoid co-cultures.
Catalog No. 50-003-93175 Supplier Medchemexpress LLC Supplier No. HY161346A5MG
(Rac)-EBET-1055 is the racemate of EBET-1055, a bromodomain and extra-terminal (BET) PROTAC degrader.
  • Effectively inhibits the growth of pancreatic ductal adenocarcinoma (PDAC).
  • Simultaneously modulates cancer-associated fibroblast (CAF) activity, upregulating all reporter gene activities in organoid co-cultures.