Learn More
TARGETMOL CHEMICALS INC RAC-EC5026 5MG
SDP
Catalog No. 502578951
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (Rac)-EC5026 ((Rac)-BPN-19186) is a potent piperidine inhibitor of soluble epoxide hydrolase (sEH) with Ki of 0.06 nM. (Rac)-EC5026 can be used for the research of Parkinson's disease and dementia with Lewy Bodies (DLB) [1]. purity: 99%
Catalog No. 50-257-8951 Supplier TARGETMOL CHEMICALS INC Supplier No. T98325MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (Rac)-EC5026 ((Rac)-BPN-19186) is a potent piperidine inhibitor of soluble epoxide hydrolase (sEH) with Ki of 0.06 nM. (Rac)-EC5026 can be used for the research of Parkinson's disease and dementia with Lewy Bodies (DLB) [1]. purity: 99%