Learn More
Medchemexpress LLC (Rac)-Etavopivat | 2622070-93-1 | 99.9% | 457.50 | 50 MG
SDP
Catalog No. 5000352114
Encompass_Preferred
(Rac)-Etavopivat, also known as (Rac)-FT-4202, is an isomer of Etavopivat. This orally active erythrocyte pyruvate kinase-R (PKR) activator is utilized in studies related to sickle cell disease and other hemoglobinopathies.
  • Improves hemoglobin-oxygen affinity and reduces the sickle point in human red blood cells.
  • Increases 2,3-DPG and ATP in crab-eating monkeys.
Catalog No. 50-003-52114 Supplier Medchemexpress LLC Supplier No. HY139573A50MG
(Rac)-Etavopivat, also known as (Rac)-FT-4202, is an isomer of Etavopivat. This orally active erythrocyte pyruvate kinase-R (PKR) activator is utilized in studies related to sickle cell disease and other hemoglobinopathies.
  • Improves hemoglobin-oxygen affinity and reduces the sickle point in human red blood cells.
  • Increases 2,3-DPG and ATP in crab-eating monkeys.