Learn More
Medchemexpress LLC (rac)-Exatecan Intermediate 1 | 102978-40-5 | 99.97% | 263.25 | 1 ML
SDP
Catalog No. 5000401610
Encompass_Preferred
(rac)-Exatecan Intermediate 1 is an isomer of Exatecan Intermediate 1 (HY-42487), which is an intermediate of Exatecan (HY-13631). Exatecan is a camptothecin-based anticancer agent that inhibits tumor growth by interfering with the proliferation and division of tumor cells by interacting with DNA. It is primarily used in research concerning various cancers.
  • Interferes with tumor cell proliferation
  • Inhibits tumor growth by interacting with DNA
  • Used in research for ovarian, lung, and breast cancer
Catalog No. 50-004-01610 Supplier Medchemexpress LLC Supplier No. HY42487A10MM/1ML
(rac)-Exatecan Intermediate 1 is an isomer of Exatecan Intermediate 1 (HY-42487), which is an intermediate of Exatecan (HY-13631). Exatecan is a camptothecin-based anticancer agent that inhibits tumor growth by interfering with the proliferation and division of tumor cells by interacting with DNA. It is primarily used in research concerning various cancers.
  • Interferes with tumor cell proliferation
  • Inhibits tumor growth by interacting with DNA
  • Used in research for ovarian, lung, and breast cancer