Learn More
Medchemexpress LLC (Rac)-Finerenone ((Rac)-BAY 94-8862) | 1050477-27-4 | 99.66% | 378.42 | 10 MG
SDP
Catalog No. 5000343358
Encompass_Preferred
(Rac)-Finerenone ((Rac)-BAY 94-8862) is the racemate of Finerenone. Finerenone is a third-generation, selective, and orally available nonsteroidal mineralocorticoid receptor (MR) antagonist with an IC50 of 18 nM. It shows excellent selectivity against glucocorticoid receptor (GR), androgen receptor (AR), and progesterone receptor (>500-fold).
  • Selective nonsteroidal mineralocorticoid receptor antagonist.
  • Orally available.
  • High selectivity against glucocorticoid, androgen, and progesterone receptors.
  • Available as a ready-to-use 10 mM solution in DMSO.
  • Good solubility in DMSO (62.5 mg/mL).
  • Multiple in vivo dissolution protocols available for experimental flexibility.
Catalog No. 50-003-43358 Supplier Medchemexpress LLC Supplier No. HY111372A10MM/1ML
(Rac)-Finerenone ((Rac)-BAY 94-8862) is the racemate of Finerenone. Finerenone is a third-generation, selective, and orally available nonsteroidal mineralocorticoid receptor (MR) antagonist with an IC50 of 18 nM. It shows excellent selectivity against glucocorticoid receptor (GR), androgen receptor (AR), and progesterone receptor (>500-fold).
  • Selective nonsteroidal mineralocorticoid receptor antagonist.
  • Orally available.
  • High selectivity against glucocorticoid, androgen, and progesterone receptors.
  • Available as a ready-to-use 10 mM solution in DMSO.
  • Good solubility in DMSO (62.5 mg/mL).
  • Multiple in vivo dissolution protocols available for experimental flexibility.