Learn More
TARGETMOL CHEMICALS INC Rac-GSK 372475 1MG
SDP
Catalog No. 502879878
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. (Rac)-GSK 372475 may be investigated for the treatment of Parkinson's syndrome, depression and obesity. Purity 96.89%
Catalog No. 50-287-9878 Supplier TARGETMOL CHEMICALS INC Supplier No. T680261MG
Also available in 5 mg, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. (Rac)-GSK 372475 may be investigated for the treatment of Parkinson's syndrome, depression and obesity. Purity 96.89%