Learn More
Medchemexpress LLC HY-114492A 5mg Medchemexpress, Rac-GSK547 CAS: Purity:>98%
SDP
Catalog No. 501932495
Encompass_Preferred
Medchemexpress, HY-114492A 5mg Rac-GSK547 CAS: Rac-GSK547 is the racemate of GSK547. GSK547 is a highly selective and potent inhibitor of receptor-interacting serine/threonine protein kinase 1 (RIP1), inhibits macrophage-mediated adaptive immune tolerance in pancreatic cancer [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2495 Supplier Medchemexpress LLC Supplier No. HY114492A5MG
Medchemexpress, HY-114492A 5mg Rac-GSK547 CAS: Rac-GSK547 is the racemate of GSK547. GSK547 is a highly selective and potent inhibitor of receptor-interacting serine/threonine protein kinase 1 (RIP1), inhibits macrophage-mediated adaptive immune tolerance in pancreatic cancer [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.