Learn More
Cayman Chemical rac-HesperetIn-d3 1mg
SDP
Catalog No. 502911105
Encompass_Preferred
An internal standard for the quantification of hesperetin by GC- or LC-MS
Catalog No. 50-291-1105 Supplier Cayman Chemical Supplier No. 358021 MG
An internal standard for the quantification of hesperetin by GC- or LC-MS