Learn More
Medchemexpress LLC (Rac)-Hexestrol-d4 | 1189950-25-1 | 1 MG
SDP
Catalog No. 5000412356
Encompass_Preferred
(Rac)-Hexestrol-d4 is the deuterium labeled (Rac)-Hexestrol. It is intended for research use only and is not sold to patients. This compound can be utilized as a tracer and as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes like deuterium into drug molecules is primarily for quantitation during the drug development process, as deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis.
  • Incorporation of stable heavy isotopes into drug molecules for quantitation.
  • Deuteration can influence pharmacokinetic and metabolic profiles.
Catalog No. 50-004-12356 Supplier Medchemexpress LLC Supplier No. HYB1662S1MG
(Rac)-Hexestrol-d4 is the deuterium labeled (Rac)-Hexestrol. It is intended for research use only and is not sold to patients. This compound can be utilized as a tracer and as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes like deuterium into drug molecules is primarily for quantitation during the drug development process, as deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis.
  • Incorporation of stable heavy isotopes into drug molecules for quantitation.
  • Deuteration can influence pharmacokinetic and metabolic profiles.