Learn More
Medchemexpress LLC (Rac)-IBT6A | 1412418-47-3 | 98.18% | 100 MG
SDP
Catalog No. 5000339861
Encompass_Preferred
(Rac)-IBT6A is a racemate of IBT6A, an impurity of Ibrutinib. This product can be used in the synthesis of IBT6A Ibrutinib dimer and IBT6A adduct. Ibrutinib is known as a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM.
  • Targets Btk
  • Functions via Protein Tyrosine Kinase/RTK pathway
  • Can be stored as powder at -20°C for 3 years or 4°C for 2 years
  • Can be stored in solvent at -80°C for 2 years or -20°C for 1 year
  • Soluble in DMSO at 50 mg/mL
  • Used in the synthesis of Ibrutinib and Ibrutinib-based activity-based probes (ABPs)
Catalog No. 50-003-39861 Supplier Medchemexpress LLC Supplier No. HY13036100MG
(Rac)-IBT6A is a racemate of IBT6A, an impurity of Ibrutinib. This product can be used in the synthesis of IBT6A Ibrutinib dimer and IBT6A adduct. Ibrutinib is known as a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM.
  • Targets Btk
  • Functions via Protein Tyrosine Kinase/RTK pathway
  • Can be stored as powder at -20°C for 3 years or 4°C for 2 years
  • Can be stored in solvent at -80°C for 2 years or -20°C for 1 year
  • Soluble in DMSO at 50 mg/mL
  • Used in the synthesis of Ibrutinib and Ibrutinib-based activity-based probes (ABPs)